Isoquinoline-5-sulfonic acid ≥97%

Supplier: Matrix Scientific

085565-10G 085565-50G
102845-382EA 181.54 USD
102845-382 102845-384
Isoquinoline-5-sulfonic acid ≥97%
Isoquinoline-5-sulphonic acid

Hazard: Irritant

Formula: C₉H₇NO₃S
MW: 209.22 g/mol
MDL Number: MFCD00134089
CAS Number: 27655-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR