2-Methylbenzimidazole ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Methyl-1H-benzo[d]imidazole, 2-Methyl-1H-benzimidazole
085551-1G
085551-10G
102845-334EA
2221.54
USD
102845-334
102845-336
2-Methylbenzimidazole ≥97%
2-Methylbenzimidazole
Hazard: Irritant
Formula:
C₈H₈N₂ MW: 132.16 g/mol Melting Pt: 176…177 °C |
MDL Number:
MFCD00005598 CAS Number: 615-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...