1-(3-Chloropropyl)-2-benzimidazolidinone ≥97%

Supplier: Matrix Scientific

085540-1G 085540-10G
102845-308EA 126.15 USD
102845-308 102845-310
1-(3-Chloropropyl)-2-benzimidazolidinone ≥97%
1-(3-Chloropropyl)-2-benzimidazolidinone

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂O
CAS Number: 62780-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR