1-Acetyl-5-nitroindoline ≥85%
Supplier: Matrix Scientific
Synonyms:
1-(5-Nitroindolin-1-yl)ethanone
085522-1G
085522-10G
085522-50G
102845-254EA
72.31
USD
102845-254
102845-256
102845-258
1-Acetyl-5-nitroindoline ≥85%
1-Acetyl-5-nitroindoline
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₃ MW: 206.2 g/mol |
MDL Number:
MFCD00005811 CAS Number: 33632-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...