3-(3-Methyl-1H-indol-1-yl)propanenitrile ≥97%

Supplier: Matrix Scientific

085519-1G 085519-10G
102845-246EA 184.62 USD
102845-246 102845-248
3-(3-Methyl-1H-indol-1-yl)propanenitrile ≥97%
3-(3-Methyl-1H-indol-1-yl)propanenitrile

Hazard: Irritant

Formula: C₁₂H₁₂N₂
MDL Number: MFCD00052087
CAS Number: 4414-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR