3-Methylbenzofuran ≥97%

Supplier: Matrix Scientific

085504-1G 085504-10G
102845-210EA 255.38 USD
102845-210 102845-212
3-Methylbenzofuran ≥97%
3-Methylbenzofuran

Hazard: Irritant

Formula: C₉H₈O
MW: 132.16 g/mol
MDL Number: MFCD00205641
CAS Number: 21535-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR