2-[(6-Nitro-1,1-dioxo-2,3-dihydro-1H-1-benzothiophen-3-yl)sulfanyl]acetohydrazide ≥97%

Supplier: Matrix Scientific

085503-1G 085503-10G
102845-206EA 184.62 USD
102845-206 102845-208
2-[(6-Nitro-1,1-dioxo-2,3-dihydro-1H-1-benzothiophen-3-yl)sulfanyl]acetohydrazide ≥97%
2-[(6-Nitro-1,1-dioxo-2,3-dihydro-1H-1-benzothiophen-3-yl)sulfanyl]acetohydrazide

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₅S₂
MDL Number: MFCD04110822
CAS Number: 685120-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR