1-Benzothiophene-2-carboxamide ≥97%

Supplier: Matrix Scientific

Synonyms: Benzo[b]thiophene-2-carboxamide

085502-1G 085502-10G
102845-202EA 101.54 USD
102845-202 102845-204
1-Benzothiophene-2-carboxamide ≥97%
1-Benzothiophene-2-carboxamide

Hazard: Irritant

Formula: C₉H₇NOS
MW: 177.23 g/mol
MDL Number: MFCD00052505
CAS Number: 6314-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR