2-(2-Bromoethoxy)tetrahydropyran ≥96%

Supplier: Matrix Scientific

Synonyms: 2-(2-Bromoethoxy)tetrahydro-2H-pyran

085486-1G 085486-10G 085486-50G
102845-160EA 156.92 USD
102845-160 102845-162 102845-164
2-(2-Bromoethoxy)tetrahydropyran ≥96%
2-(2-Bromoethoxy)tetrahydropyran

Hazard: Irritant

Formula: C₇H₁₃BrO₂
MW: 209.0809 g/mol
Boiling Pt: 75°C/1 Torr
MDL Number: MFCD01321310
CAS Number: 17739-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR