2-(3-Bromopropoxy)tetrahydropyran ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(3-Bromopropoxy)tetrahydro-2H-pyran

085476-1G 085476-5G
102845-132EA 69.23 USD
102845-132 102845-134
2-(3-Bromopropoxy)tetrahydropyran ≥97%
2-(3-Bromopropoxy)tetrahydropyran

Hazard: Irritant

Formula: C₈H₁₅BrO₂
MDL Number: MFCD00058593
CAS Number: 33821-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR