2-Piperazin-1-yl-4-(trifluoromethyl)pyrimidine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Piperazin-1-yl)-4-(trifluoromethyl)pyrimidine

085434-1G 085434-10G
102845-042EA 283.08 USD
102845-042 102845-044
2-Piperazin-1-yl-4-(trifluoromethyl)pyrimidine ≥97%
2-Piperazin-1-yl-4-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₉H₁₁F₃N₄
MW: 232.21 g/mol
MDL Number: MFCD00203912
CAS Number: 179756-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR