5-Phenylhydantoin ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Phenylimidazolidine-2,4-dione
085410-1G
085410-10G
085410-50G
102844-976EA
101.54
USD
102844-976
102844-978
102844-980
5-Phenylhydantoin ≥97%
5-Phenylhydantoin
Hazard: Irritant
Formula:
C₉H₈N₂O₂ MW: 176.1720 g/mol |
MDL Number:
MFCD00037881 CAS Number: 89-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...