1-(2-Chloroethyl)imidazole hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(2-Chloroethyl)-1H-imidazole hydrochloride

085405-1G 085405-10G
102844-960EA 101.54 USD
102844-960 102844-962
1-(2-Chloroethyl)imidazole hydrochloride ≥97%
1-(2-Chloroethyl)imidazole hydrochloride

Hazard: Irritant

Formula: C₅H₈Cl₂N₂
MW: 167.04 g/mol
MDL Number: MFCD00799332
CAS Number: 18994-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR