1-Benzylimidazole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Benzyl-1H-imidazole

085404-1G 085404-10G
102844-956EA 156.92 USD
102844-956 102844-958
1-Benzylimidazole ≥97%
1-Benzylimidazole

Hazard: Irritant

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 310
Melting Pt: 72
MDL Number: MFCD00005296
CAS Number: 4238-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR