2-(4,5-Dichloro-1H-imidazol-1-yl)ethanethioamide ≥97%

Supplier: Matrix Scientific

085403-1G 085403-10G
102844-952EA 127.69 USD
102844-952 102844-954
2-(4,5-Dichloro-1H-imidazol-1-yl)ethanethioamide ≥97%
2-(4,5-Dichloro-1H-imidazol-1-yl)ethanethioamide

Hazard: Irritant

Formula: C₅H₅Cl₂N₃S
MW: 210.09 g/mol
MDL Number: MFCD00084996
CAS Number: 175201-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR