5-(4-Chlorobutyl)hydantoin ≥97%

Supplier: Matrix Scientific

085400-1G 085400-10G
102844-942EA 101.54 USD
102844-942 102844-944
5-(4-Chlorobutyl)hydantoin ≥97%
5-(4-Chlorobutyl)hydantoin

Hazard: Irritant

Formula: C₇H₁₁ClN₂O₂
MDL Number: MFCD00059722
CAS Number: 40126-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR