4-tert-Butyl-2-methylthiazole ≥97%

Supplier: Matrix Scientific

085389-1G 085389-10G
102844-918EA 127.69 USD
102844-918 102844-920
4-tert-Butyl-2-methylthiazole ≥97%
4-tert-Butyl-2-methylthiazole

Purity: 97% CAS: 15679-11-5 MDL Number: MFCD00051643 Molecular Formula: C8H13NS Molecular Weight: 155,26 Hazard: Irritant

Formula: C₈H₁₃NS
MW: 155.26 g/mol
MDL Number: MFCD00051643
CAS Number: 15679-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR