5-(4-Fluorophenyl)oxazole ≥97%

Supplier: Matrix Scientific

085352-1G 085352-10G
102844-828EA 200 USD
102844-828 102844-830
5-(4-Fluorophenyl)oxazole ≥97%
5-(4-Fluorophenyl)oxazole

Hazard: Irritant

Formula: C₉H₆FNO
MW: 163.1485 g/mol
MDL Number: MFCD00085087
CAS Number: 128101-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR