3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxamide ≥97%

Supplier: Matrix Scientific

085350-1G 085350-10G
102844-822EA 270.77 USD
102844-822 102844-824
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxamide ≥97%
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxamide

Hazard: Irritant

Formula: C₁₁H₈Cl₂N₂O₂
MDL Number: MFCD00052211
CAS Number: 23858-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR