2-(5-Isoxazolyl)phenol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Isoxazol-5-yl)phenol

085349-1G 085349-10G
102844-818EA 101.54 USD
102844-818 102844-820
2-(5-Isoxazolyl)phenol ≥97%
2-(5-Isoxazolyl)phenol

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD00218653
CAS Number: 61348-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR