4-[(5-Methylisoxazol-3-yl)amino]-4-oxobut-2-enoic acid ≥97%

Supplier: Matrix Scientific

085346-1G 085346-10G
102844-810EA 127.69 USD
102844-810 102844-812
4-[(5-Methylisoxazol-3-yl)amino]-4-oxobut-2-enoic acid ≥97%
4-[(5-Methylisoxazol-3-yl)amino]-4-oxobut-2-enoic acid

Hazard: Irritant

Formula: C₈H₈N₂O₄
MDL Number: MFCD00175047
CAS Number: 306935-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR