3-Hydroxy-5-methylisoxazole ≥97%

Supplier: Matrix Scientific

Synonyms: Hymexazol

085339-1G 085339-10G 085339-50G
102844-788EA 26.15 USD
102844-788 102844-790 102844-792
3-Hydroxy-5-methylisoxazole ≥97%
3-Hydroxy-5-methylisoxazole

Hazard: Irritant

Formula: C₄H₅NO₂
MW: 99.09 g/mol
MDL Number: MFCD00144468
CAS Number: 10004-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR