1,3-Thiazolin-4-one-2-acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: (4,5-Dihydro-4-oxo-1,3-thiazol-2-yl)acetonitrile

085331-1G 085331-10G
102844-768EA 127.69 USD
102844-768 102844-770
1,3-Thiazolin-4-one-2-acetonitrile ≥95%
1,3-Thiazolin-4-one-2-acetonitrile

Hazard: Irritant

Formula: C₅H₄N₂OS
MW: 140.16 g/mol
MDL Number: MFCD00052566
CAS Number: 74246-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR