N-(4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride ≥97%

Supplier: Matrix Scientific

085329-1G 085329-10G
102844-760EA 103.08 USD
102844-760 102844-762
N-(4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride ≥97%
N-(4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride

Hazard: Irritant

Formula: C₅H₉ClN₄S
MDL Number: MFCD00053024
CAS Number: 100599-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR