N-Phenylmaleimide ≥97%

Supplier: Matrix Scientific

085321-1G 085321-10G 085321-50G
102844-738EA 46.15 USD
102844-738 102844-740 102844-742
N-Phenylmaleimide ≥97%
N-Phenylmaleimide

Hazard: Irritant

Formula: C₁₀H₇NO₂
MDL Number: MFCD00005502
CAS Number: 941-69-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR