3-Methyl-2(5H)-furanone ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-2-methyl-2-butenoic acid gamma-Lactone

085299-1G 085299-10G
102844-690EA 270.77 USD
102844-690 102844-692
3-Methyl-2(5H)-furanone ≥97%
3-Methyl-2(5H)-furanone

Hazard: Irritant

Formula: C₅H₆O₂
MW: 98.1 g/mol
MDL Number: MFCD00191545
CAS Number: 22122-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR