3-(2-Methyl-3-furyl)-3-oxopropanenitrile ≥97%

Supplier: Matrix Scientific

085295-1G 085295-10G
102844-676EA 143.08 USD
102844-676 102844-678
3-(2-Methyl-3-furyl)-3-oxopropanenitrile ≥97%
3-(2-Methyl-3-furyl)-3-oxopropanenitrile

Hazard: Irritant

Formula: C₈H₇NO₂
MDL Number: MFCD00068045
CAS Number: 158386-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR