2,5-Dimethyl-3(2H)-furanone ≥97%

Supplier: Matrix Scientific

Synonyms: 2,5-Dimethylfuran-3(2H)-one

085292-10G 085292-50G
102844-668EA 387.69 USD
102844-668 102844-670
2,5-Dimethyl-3(2H)-furanone ≥97%
2,5-Dimethyl-3(2H)-furanone

Hazard: Irritant

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00052571
CAS Number: 14400-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR