2-Chloro-1-(4,5-dichloro-2-thienyl)ethan-1-one ≥85%
Supplier: Matrix Scientific
085288-1G
085288-10G
102844-656EA
553.85
USD
102844-656
102844-658
2-Chloro-1-(4,5-dichloro-2-thienyl)ethan-1-one ≥85%
2-Chloro-1-(4,5-dichloro-2-thienyl)ethan-1-one
Hazard: Irritant
Formula:
C₆H₃Cl₃OS |
MDL Number:
MFCD02090184 CAS Number: 64218-50-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...