2-Amino-4-methylthiophene-3-carboxamide ≥97%
Supplier: Matrix Scientific
085271-1G
085271-10G
102844-622EA
156.92
USD
102844-622
102844-624
2-Amino-4-methylthiophene-3-carboxamide ≥97%
2-Amino-4-methylthiophene-3-carboxamide
Hazard: Irritant
Formula:
C₆H₈N₂OS MW: 156.20 g/mol |
MDL Number:
MFCD00052594 CAS Number: 4651-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...