3-Methyl-2-thiophenecarbonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Methylthiophene-2-carbonitrile

085269-1G 085269-10G
102844-616EA 46.15 USD
102844-616 102844-618
3-Methyl-2-thiophenecarbonitrile ≥97%
3-Methyl-2-thiophenecarbonitrile

Hazard: Toxic

Formula: C₆H₅NS
MW: 123.18 g/mol
Boiling Pt: 91-93°C/12mm
MDL Number: MFCD00052086
CAS Number: 55406-13-8
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR