Thiophen-2-carbothioamide ≥97%

Supplier: Matrix Scientific

Synonyms: Thiophene-2-carbothioamide

085267-1G 085267-10G
102844-610EA 184.62 USD
102844-610 102844-612
Thiophen-2-carbothioamide ≥97%
Thiophen-2-carbothioamide

Hazard: Irritant

Formula: C₅H₅NS₂
MW: 143.23 g/mol
MDL Number: MFCD00052890
CAS Number: 20300-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR