Oxo(thien-2-yl)acetonitrile ≥97%

Supplier: Matrix Scientific

085254-1G 085254-10G
102844-580EA 383.08 USD
102844-580 102844-582
Oxo(thien-2-yl)acetonitrile ≥97%
Oxo(thien-2-yl)acetonitrile

Hazard: Irritant

Formula: C₆H₃NOS
MDL Number: MFCD01935213
CAS Number: 6007-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR