2,6-Di-tert-butyl-4-methylpyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 2,6-Di-Tert-butyl-4-methylpyridine

085238-5G 085238-25G
102844-538EA 80 USD
102844-538 102844-540
2,6-Di-tert-butyl-4-methylpyridine ≥97%
2,6-Di-tert-butyl-4-methylpyridine

Hazard: Irritant/Light Sensitive/Air Sensitive

Formula: C₁₄H₂₃N
Boiling Pt: 232…234 °C
Melting Pt: 30…35 °C
Flash Pt: 83 °C
MDL Number: MFCD00006305
CAS Number: 38222-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR