2,2'-Dithiobis(5-nitropyridine) ≥97%

Supplier: Matrix Scientific

Synonyms: 1,2-Bis(5-nitropyridin-2-yl)disulphane

085226-1G 085226-10G 085226-50G
102844-510EA 123.08 USD
102844-510 102844-512 102844-514
2,2'-Dithiobis(5-nitropyridine) ≥97%
2,2'-Dithiobis(5-nitropyridine)

Hazard: Irritant

Formula: C₁₀H₆N₄O₄S₂
MW: 310.31 g/mol
Melting Pt: 150…151 °C
MDL Number: MFCD00006453
CAS Number: 2127-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR