2-(4-Methoxyphenoxy)-3-nitropyridine ≥97%

Supplier: Matrix Scientific

085220-1G 085220-10G
102844-494EA 156.92 USD
102844-494 102844-496
2-(4-Methoxyphenoxy)-3-nitropyridine ≥97%
2-(4-Methoxyphenoxy)-3-nitropyridine

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₄
MDL Number: MFCD00829021
CAS Number: 76893-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR