2-(4-Chlorophenoxy)-3-nitropyridine ≥97%

Supplier: Matrix Scientific

085219-1G 085219-10G
102844-490EA 156.92 USD
102844-490 102844-492
2-(4-Chlorophenoxy)-3-nitropyridine ≥97%
2-(4-Chlorophenoxy)-3-nitropyridine

Hazard: Irritant

Formula: C₁₁H₇ClN₂O₃
CAS Number: 76893-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR