Methyl-2-{[5-(trifluoromethyl)-2-pyridyl]sulfonyl}acetate ≥97%

Supplier: Matrix Scientific

085218-1G 085218-10G
102844-486EA 298.46 USD
102844-486 102844-488
Methyl-2-{[5-(trifluoromethyl)-2-pyridyl]sulfonyl}acetate ≥97%
Methyl-2-{[5-(trifluoromethyl)-2-pyridyl]sulfonyl}acetate

Hazard: Irritant

Formula: C₉H₈F₃NO₄S
MDL Number: MFCD00107950
CAS Number: 142274-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR