2-(4-Chlorophenoxy)-5-nitropyridine ≥97%

Supplier: Matrix Scientific

085215-1G 085215-10G
102844-476EA 241.54 USD
102844-476 102844-478
2-(4-Chlorophenoxy)-5-nitropyridine ≥97%
2-(4-Chlorophenoxy)-5-nitropyridine

Hazard: Irritant

Formula: C₁₁H₇ClN₂O₃
CAS Number: 28232-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR