3-Hydroxy-2-nitro-6-picoline ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Methyl-2-nitropyridin-3-ol, 6-Methyl-2-nitro-3-pyridone, 3-Hydroxy-6-methyl-2-nitropyridine, 6-Metil-2-nitropyridine-3-ol

085153-1G 085153-10G
102844-344EA 101.54 USD
102844-344 102844-346
3-Hydroxy-2-nitro-6-picoline ≥97%
3-Hydroxy-2-nitro-6-picoline

Hazard: Irritant

Formula: C₆H₆N₂O₃
MDL Number: MFCD00006259
CAS Number: 15128-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR