2-Ethoxy-5-nitropyridine ≥97%

Supplier: Matrix Scientific

085143-1G 085143-10G 085143-50G
102844-320EA 56.92 USD
102844-320 102844-322 102844-324
2-Ethoxy-5-nitropyridine ≥97%
2-Ethoxy-5-nitropyridine

Hazard: Irritant

Formula: C₇H₈N₂O₃
MDL Number: MFCD01646181
CAS Number: 31594-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR