3-Ethoxy-2-nitropyridine ≥97%

Supplier: Matrix Scientific

085137-1G 085137-10G
102844-304EA 156.92 USD
102844-304 102844-306
3-Ethoxy-2-nitropyridine ≥97%
3-Ethoxy-2-nitropyridine

Hazard: Irritant

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
MDL Number: MFCD00006257
CAS Number: 74037-50-6
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR