2,4-Dimethyl-3-nitropyridine ≥97%

Supplier: Matrix Scientific

085115-5G 085115-25G
102844-256EA 143.08 USD
102844-256 102844-258
2,4-Dimethyl-3-nitropyridine ≥97%
2,4-Dimethyl-3-nitropyridine

Hazard: Irritant

Formula: C₇H₈N₂O₂
MDL Number: MFCD08235269
CAS Number: 1074-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR