1,3-Di(2-pyridyl)-1,3-propanedione ≥97%
Supplier: Matrix Scientific
Synonyms:
Dipicolinoylmethane, 1,3-Di(pyridin-2-yl)propane-1,3-dione
085103-1G
085103-10G
102844-226EA
241.54
USD
102844-226
102844-228
1,3-Di(2-pyridyl)-1,3-propanedione ≥97%
1,3-Di(2-pyridyl)-1,3-propanedione
Hazard: Irritant
Formula:
C₁₃H₁₀N₂O₂ |
MDL Number:
MFCD01321157 CAS Number: 10198-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...