[(2-Chloroethoxy)(phenyl)methyl]benzene ≥97%

Supplier: Matrix Scientific

084071-1G 084071-10G
102841-446EA 298.46 USD
102841-446 102841-448
[(2-Chloroethoxy)(phenyl)methyl]benzene ≥97%
[(2-Chloroethoxy)(phenyl)methyl]benzene

Hazard: Irritant

Formula: C₁₅H₁₅ClO
Melting Pt: 30…31 °C
MDL Number: MFCD00037115
CAS Number: 32669-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR