6-Iodo-1,4-benzodioxane ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Iodo-2,3-dihydro-1,4-benzodioxine, 1,2-Ethylenedioxy-4-iodobenzene, 6-Iodo-2,3-dihydrobenzo[b][1,4]dioxine

084045-1G 084045-10G 084045-50G
102841-386EA 72.31 USD
102841-386 102841-388 102841-390
6-Iodo-1,4-benzodioxane ≥95%
6-Iodo-1,4-benzodioxane

Hazard: Irritant

Formula: C₈H₇IO₂
MW: 262.05 g/mol
MDL Number: MFCD00221460
CAS Number: 57744-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR