7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine ≥97%

Supplier: Matrix Scientific

Synonyms: 7-Bromo-4-methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine, 7-Bromo-3,4-dihydro-4-methyl-2H-1,4-benzoxazine

084044-1G 084044-10G
102841-382EA 524.62 USD
102841-382 102841-384
7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine ≥97%
7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine

Hazard: Irritant

Formula: C₉H₁₀BrNO
MW: 228.1 g/mol
MDL Number: MFCD02681913
CAS Number: 154264-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR