4-Chloroquinaldine ≥97%

Supplier: Matrix Scientific

084025-1G 084025-10G 084025-50G
102841-340EA 56.92 USD
102841-340 102841-342 102841-344
4-Chloroquinaldine ≥97%
4-Chloroquinaldine

Hazard: Irritant

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Boiling Pt: 269 °C
Melting Pt: 42 °C
MDL Number: MFCD00006757
CAS Number: 4295-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR