5-Chloro-2,3,3-trimethyl-3H-indole ≥97%
Supplier: Matrix Scientific
084005-1G
084005-10G
102841-284EA
241.54
USD
102841-284
102841-286
5-Chloro-2,3,3-trimethyl-3H-indole ≥97%
5-Chloro-2,3,3-trimethyl-3H-indole
Hazard: Irritant
Formula:
C₁₁H₁₂ClN MW: 193.68 g/mol |
MDL Number:
MFCD00126966 CAS Number: 25981-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...