1-Boc-5-bromoindole ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-5-bromoindole, 5-Bromo-1-tert-butoxycarbonylindole, tert-Butyl 5-bromo-1H-indole-1-carboxylate

084004-1G 084004-10G
102841-280EA 127.69 USD
102841-280 102841-282
1-Boc-5-bromoindole ≥95%
1-Boc-5-bromoindole

Hazard: Irritant

Formula: C₁₃H₁₄BrNO₂
MW: 296.16
MDL Number: MFCD02090067
CAS Number: 182344-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR